Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017396.2 | NP_001017396.1 | 4 | 1380 | ||
NM_001282398.1 | NP_001269327.1 | 4 | 1338 | |||
NM_001291604.1 | NP_001278533.1 | 3 | 1266 | |||
NM_001291605.1 | NP_001278534.1 | 5 | 1491 | |||
NM_021088.3 | NP_066574.2 | 5 | 1452 | |||
GenBank mRNA | AK055072.1 | - | 5 | 1432 | ||
AK290798.1 | - | 4 | 990 | |||
AK295739.1 | - | 4 | 969 | |||
AK299058.1 | - | 5 | 1091 | |||
AK315762.1 | - | 5 | 1064 | |||
AK315829.1 | - | 4 | 985 | |||
BC005068.1 | - | 5 | 1424 | 3350363 | ||
BC009976.2 | - | 3 | 838 | |||
CR749650.1 | - | 3 | 1217 | |||
X60152.1 | - | 4 | 1365 |