Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IKZF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022465.3 | NP_071910.3 | 6 | 1364 | ||
XM_005269086.3 | XP_005269143.1 | 8 | 906 | |||
XM_005269089.2 | XP_005269146.1 | 6 | 1008 | |||
XM_005269090.3 | XP_005269147.1 | 7 | 812 | |||
XM_005269093.2 | XP_005269150.1 | 5 | 1283 | |||
XM_011538664.1 | XP_011536966.1 | 8 | 909 | |||
XM_011538666.2 | XP_011536968.1 | 7 | 757 | |||
XM_011538669.2 | XP_011536971.1 | 6 | 707 | |||
XM_017019805.1 | XP_016875294.1 | 10 | 1154 | |||
XM_017019806.1 | XP_016875295.1 | 10 | 1397 | |||
XM_017019807.1 | XP_016875296.1 | 10 | 1394 | |||
XM_017019808.1 | XP_016875297.1 | 9 | 1079 | |||
XM_017019809.1 | XP_016875298.1 | 10 | 1151 | |||
XM_017019810.1 | XP_016875299.1 | 10 | 1249 | |||
XM_017019811.1 | XP_016875300.1 | 9 | 1076 | |||
XM_017019812.1 | XP_016875301.1 | 8 | 1152 | |||
XM_017019813.1 | XP_016875302.1 | 8 | 1149 | |||
XM_017019814.1 | XP_016875303.1 | 7 | 1058 | |||
XM_017019815.1 | XP_016875304.1 | 7 | 1055 | |||
XM_017019816.1 | XP_016875305.1 | 7 | 815 | |||
GenBank mRNA | AB058685.1 | - | 4 | 908 | ||
AF230809.1 | - | 5 | 616 | |||
AK122880.1 | - | 5 | 1131 | |||
AK314543.1 | - | 5 | 729 | |||
BX647761.1 | - | 7 | 934 |