Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001755.2 | NP_001746.1 | 5 | 720 | ||
NM_022845.2 | NP_074036.1 | 5 | 720 | |||
XM_005256212.3 | XP_005256269.1 | 4 | 565 | |||
XM_006721321.2 | XP_006721384.1 | 4 | 565 | |||
XM_017023812.1 | XP_016879301.1 | 4 | 565 | |||
XM_017023813.1 | XP_016879302.1 | 4 | 589 | |||
GenBank mRNA | AF294326.1 | - | 5 | 459 | ||
AK290462.1 | - | 5 | 478 | |||
AK291834.1 | - | 5 | 462 | |||
AK311084.1 | - | 1 | 47 | |||
BC018509.1 | - | 5 | 720 | 3867061 | ||
BT006862.1 | - | 5 | 459 | |||
L20298.1 | - | 5 | 438 |