Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF7L2 SIRNA Assays ›
Gene Symbol:
TCF7L2
Gene Name:
transcription factor 7 like 2
Gene Aliases:
TCF-4, TCF4
Chromosome Location:
Chr.10: 112950219 - 113167678 on Build GRCh38
UniGene ID:
Hs.593995
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
116411
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF7L2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146274.1
NP_001139746.1
5, 6
1053
NM_001146284.1
NP_001139756.1
4, 5
984
NM_001146285.1
NP_001139757.1
4, 5
984
NM_001146286.1
NP_001139758.1
4, 5
984
NM_001198525.1
NP_001185454.1
4, 5
984
NM_001198526.1
NP_001185455.1
4, 5
984
NM_001198527.1
NP_001185456.1
4, 5
984
NM_001198528.1
NP_001185457.1
4, 5
984
NM_001198529.1
NP_001185458.1
4, 5
984
NM_001198531.1
NP_001185460.1
5, 6
1053
NM_030756.4
NP_110383.2
4, 5
984
XM_005270084.1
XP_005270141.1
5, 6
1084
XM_005270085.1
XP_005270142.1
5, 6
1084
XM_005270086.1
XP_005270143.1
4, 5
1015
XM_005270088.1
XP_005270145.1
4, 5
1015
XM_005270089.1
XP_005270146.1
4, 5
1015
XM_005270096.2
XP_005270153.1
5, 6
1084
XM_005270100.1
XP_005270157.1
5, 6
1084
XM_005270101.2
XP_005270158.1
4, 5
1015
XM_005270102.1
XP_005270159.1
5, 6
1084
XM_005270103.1
XP_005270160.1
4, 5
1015
XM_005270104.1
XP_005270161.1
4, 5
1015
XM_011540111.1
XP_011538413.1
4, 5
1015
XM_017016590.1
XP_016872079.1
5, 6
1084
XM_017016591.1
XP_016872080.1
5, 6
1084
XM_017016592.1
XP_016872081.1
5, 6
1084
XM_017016593.1
XP_016872082.1
5, 6
1084
XM_017016594.1
XP_016872083.1
4, 5
1015
XM_017016596.1
XP_016872085.1
5, 6
1084
GenBank mRNA
AB440195.1
-
5, 6
546
AB451266.1
-
4, 5
477
AK225809.1
-
4, 5
988
AK299295.1
-
4, 5
514
BC032656.1
-
4, 5
573
5533185
BX648364.1
-
5, 6
421
CR536574.1
-
4, 5
477
FJ010164.1
-
3, 4
372
FJ010166.1
-
4, 5
570
FJ010167.1
-
4, 5
570
FJ010168.1
-
4, 5
574
FJ010169.1
-
4, 5
574
FJ010170.1
-
4, 5
570
FJ010172.1
-
4, 5
574
FJ010173.1
-
3, 4
372
FJ010174.1
-
4, 5
932
HM352838.1
-
4, 5
477
HM352839.1
-
4, 5
477
HM352840.1
-
5, 6
546
HM352841.1
-
5, 6
546
HM352842.1
-
4, 5
477
HM352843.1
-
4, 5
477
HM352844.1
-
5, 6
546
HM352846.1
-
5, 6
546
HM352847.1
-
4, 5
477
HM352848.1
-
4, 5
477
HM352849.1
-
4, 5
477
HM352850.1
-
5, 6
546
HM352851.1
-
5, 6
546
Y11306.2
-
4, 5
784
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
blood vessel development
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cell cycle arrest
Wnt signaling pathway, calcium modulating pathway
cell proliferation
response to glucose
positive regulation of heparan sulfate proteoglycan biosynthetic process
pancreas development
positive regulation of insulin secretion
positive regulation of protein binding
regulation of hormone metabolic process
glucose homeostasis
negative regulation of sequence-specific DNA binding transcription factor activity
maintenance of DNA repeat elements
canonical Wnt signaling pathway involved in positive regulation of epithelial to mesenchymal transition
fat cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein export from nucleus
myoblast fate commitment
regulation of smooth muscle cell proliferation
positive regulation of protein kinase B signaling
canonical Wnt signaling pathway
negative regulation of canonical Wnt signaling pathway
beta-catenin-TCF complex assembly
negative regulation of type B pancreatic cell apoptotic process
negative regulation of extrinsic apoptotic signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II repressing transcription factor binding
transcription factor activity, sequence-specific DNA binding
protein binding
beta-catenin binding
transcription factor binding
protein kinase binding
nuclear hormone receptor binding
sequence-specific DNA binding
transcription regulatory region DNA binding
gamma-catenin binding
armadillo repeat domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0