Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ODF2 SIRNA Assays ›
Gene Symbol:
ODF2
Gene Name:
outer dense fiber of sperm tails 2
Gene Aliases:
CT134, ODF2/1, ODF2/2, ODF84
Chromosome Location:
Chr.9: 128455155 - 128501292 on Build GRCh38
UniGene ID:
Hs.129055
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
11642
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ODF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242352.1
NP_001229281.1
5
726
NM_001242353.1
NP_001229282.1
5
812
NM_002540.4
NP_002531.3
7
1014
NM_153432.1
NP_702910.1
4
572
NM_153433.1
NP_702911.1
5
739
NM_153435.1
NP_702913.1
5
632
NM_153436.1
NP_702914.1
4
396
NM_153437.2
NP_702915.1
5
662
NM_153439.1
NP_702917.1
4
515
NM_153440.1
NP_702918.1
4
442
XM_005252010.2
XP_005252067.1
4
524
XM_005252011.2
XP_005252068.1
4
464
XM_005252012.3
XP_005252069.1
5
714
XM_005252013.3
XP_005252070.1
5
661
XM_005252014.3
XP_005252071.1
5
710
XM_005252017.4
XP_005252074.1
5
615
XM_005252018.2
XP_005252075.1
6
820
XM_005252019.3
XP_005252076.1
6
839
XM_005252022.2
XP_005252079.1
5
638
XM_005252023.2
XP_005252080.1
5
609
XM_005252024.3
XP_005252081.1
5
628
XM_005252026.4
XP_005252083.1
5
557
XM_005252029.2
XP_005252086.1
5
638
XM_006717126.3
XP_006717189.1
5
714
XM_011518722.2
XP_011517024.1
5
638
XM_011518723.2
XP_011517025.1
5
581
XM_011518724.2
XP_011517026.1
6
802
XM_011518725.2
XP_011517027.1
6
806
XM_011518726.2
XP_011517028.1
6
824
XM_011518728.2
XP_011517030.1
5
575
XM_011518729.2
XP_011517031.1
5
511
XM_011518730.2
XP_011517032.1
5
638
XM_011518732.2
XP_011517034.1
5
424
XM_011518744.1
XP_011517046.1
4
464
XM_017014764.1
XP_016870253.1
6
795
XM_017014765.1
XP_016870254.1
6
814
XM_017014766.1
XP_016870255.1
4
524
XM_017014767.1
XP_016870256.1
6
806
XM_017014768.1
XP_016870257.1
5
598
XM_017014769.1
XP_016870258.1
5
627
XM_017014770.1
XP_016870259.1
6
782
XM_017014771.1
XP_016870260.1
6
763
XM_017014772.1
XP_016870261.1
6
839
XM_017014773.1
XP_016870262.1
6
815
XM_017014774.1
XP_016870263.1
5
525
XM_017014775.1
XP_016870264.1
6
782
XM_017014776.1
XP_016870265.1
6
763
XM_017014777.1
XP_016870266.1
5
565
GenBank mRNA
AF012549.1
-
5
602
AF053970.1
-
3
301
AK299303.1
-
5
574
AK301754.1
-
4
439
AK301842.1
-
4
364
AK301855.1
-
4
418
AK302684.1
-
5
666
AK309917.1
-
5
517
AY319414.1
-
4
548
AY366499.1
-
5
509
BC010629.1
-
7
812
3874501
BC091500.1
-
5
626
30378678
BI831601.1
-
5
653
BM563800.1
-
496
DB044506.1
-
4
499
DC397869.1
-
4
396
DQ444713.1
-
3
339
DQ444714.1
-
3
282
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001113214, NM_013615, NM_001177659, NM_001177661, NM_001113213, and others.
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
multicellular organism development
spermatid development
structural molecule activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0