Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138352.1 | NP_612361.1 | 977 | |||
GenBank mRNA | AY453840.1 | - | 977 | |||
BC007384.2 | - | 2 | 71 | |||
BC030129.2 | - | 2 | 250 | |||
BC065477.1 | - | 2 | 218 | |||
BC080588.1 | - | 2 | 71 |