Gene Symbol: PARN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134477.2 | NP_001127949.1 | 2, 3 | 168 | ||
NM_001242992.1 | NP_001229921.1 | 2, 3 | 251 | |||
NM_002582.3 | NP_002573.1 | 2, 3 | 251 | |||
XM_011522510.2 | XP_011520812.1 | 2, 3 | 257 | |||
XM_011522511.2 | XP_011520813.1 | 2, 3 | 256 | |||
XM_011522513.2 | XP_011520815.1 | 2, 3 | 167 | |||
XM_011522514.1 | XP_011520816.1 | 2, 3 | 255 | |||
XM_017023258.1 | XP_016878747.1 | 2, 3 | 255 | |||
GenBank mRNA | AJ005698.1 | - | 2, 3 | 162 | ||
AK293189.1 | - | 2, 3 | 160 | |||
AK299653.1 | - | 2, 3 | 218 | |||
AK301648.1 | - | 2, 3 | 241 | |||
AK303348.1 | - | 2, 3 | 198 | |||
AK315020.1 | - | 2, 3 | 247 | |||
BC044763.1 | - | 2, 3 | 189 | |||
BC050029.1 | - | 2, 3 | 189 | 4825218 | ||
DA308295.1 | - | 2, 3 | 253 |