Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023560.3 | NP_001018854.2 | 4 | 845 | ||
NM_001111039.2 | NP_001104509.1 | 4 | 842 | |||
NM_001287421.1 | NP_001274350.1 | 4 | 331 | |||
NM_001287422.1 | NP_001274351.1 | 2 | 241 | |||
NM_152736.5 | NP_689949.3 | 4 | 440 | |||
XM_011514862.2 | XP_011513164.1 | 4 | 1044 | |||
XM_011514864.1 | XP_011513166.1 | 5 | 621 | |||
XM_011514866.1 | XP_011513168.1 | 4 | 393 | |||
XM_011514867.1 | XP_011513169.1 | 4 | 757 | |||
XM_011514869.1 | XP_011513171.1 | 5 | 685 | |||
XM_017011264.1 | XP_016866753.1 | 4 | 1042 | |||
XM_017011265.1 | XP_016866754.1 | 4 | 776 | |||
GenBank mRNA | AK095948.1 | - | 4 | 392 | ||
AK293524.1 | - | 4 | 565 | |||
AK295393.1 | - | 3 | 995 | |||
AK297613.1 | - | 2 | 102 | |||
AK301081.1 | - | 4 | 736 | |||
AL526328.3 | - | 4 | 697 | |||
AL832741.1 | - | 3 | 1033 | |||
BC013951.2 | - | 4 | 296 | 2963683 | ||
BC013962.2 | - | 4 | 296 | 2963683 | ||
BX537535.1 | - | 4 | 713 | |||
DA167287.1 | - | 4 | 329 | |||
DA341149.1 | - | 4 | 329 | |||
DA528922.1 | - | 2 | 102 | |||
DA566660.1 | - | 4 | 441 | |||
DB262147.1 | - | 4 | 307 | |||
Z11773.1 | - | 3 | 362 |