Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171815.1 | NP_001165286.1 | 5 | 1007 | ||
NM_001171816.1 | NP_001165287.1 | 6 | 1856 | |||
NM_178841.3 | NP_849163.1 | 6 | 1279 | |||
XM_011522845.2 | XP_011521147.1 | 8 | 1612 | |||
XM_011522846.1 | XP_011521148.1 | 7 | 1461 | |||
XM_011522847.1 | XP_011521149.1 | 6 | 1213 | |||
XM_017022910.1 | XP_016878399.1 | 6 | 5254 | |||
XM_017022913.1 | XP_016878402.1 | 6 | 2510 | |||
GenBank mRNA | AK026860.1 | - | 1 | 35 | ||
AK057106.1 | - | 6 | 1856 | |||
AK057201.1 | - | 5 | 927 | |||
BC013948.2 | - | 6 | 1205 | 3509621 | ||
BC017226.1 | - | 6 | 1257 | 4299817 |