Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HKR1 SIRNA Assays ›
Gene Symbol:
HKR1
Gene Name:
HKR1, GLI-Kruppel zinc finger family member
Gene Aliases:
ZNF875
Chromosome Location:
Chr.19: 37317900 - 37364455 on Build GRCh38
UniGene ID:
Hs.657027
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
116739
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HKR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_181786.2
NP_861451.1
6
2673
XM_017026672.1
XP_016882161.1
4
2861
XM_017026673.1
XP_016882162.1
4
2927
XM_017026674.1
XP_016882163.1
8
3227
XM_017026675.1
XP_016882164.1
8
3224
XM_017026676.1
XP_016882165.1
5
2522
XM_017026677.1
XP_016882166.1
5
2509
XM_017026678.1
XP_016882167.1
5
2519
XM_017026679.1
XP_016882168.1
5
2506
XM_017026680.1
XP_016882169.1
8
2889
XM_017026681.1
XP_016882170.1
7
2837
XM_017026682.1
XP_016882171.1
8
2886
XM_017026683.1
XP_016882172.1
7
2748
XM_017026684.1
XP_016882173.1
6
2610
XM_017026685.1
XP_016882174.1
4
2455
XM_017026686.1
XP_016882175.1
7
2745
XM_017026687.1
XP_016882176.1
6
2607
XM_017026688.1
XP_016882177.1
4
4679
XM_017026689.1
XP_016882178.1
6
2693
XM_017026690.1
XP_016882179.1
4
2473
XM_017026691.1
XP_016882180.1
3
2253
XM_017026692.1
XP_016882181.1
8
2823
XM_017026693.1
XP_016882182.1
9
2944
XM_017026694.1
XP_016882183.1
9
2894
XM_017026695.1
XP_016882184.1
8
2866
XM_017026696.1
XP_016882185.1
7
2718
XM_017026697.1
XP_016882186.1
6
2640
XM_017026698.1
XP_016882187.1
6
2618
XM_017026699.1
XP_016882188.1
4
5454
XM_017026700.1
XP_016882189.1
6
2619
XM_017026701.1
XP_016882190.1
8
2817
XM_017026702.1
XP_016882191.1
7
2739
XM_017026703.1
XP_016882192.1
5
2516
XM_017026704.1
XP_016882193.1
5
2578
XM_017026705.1
XP_016882194.1
2
2575
XM_017026707.1
XP_016882196.1
5
2991
XM_017026708.1
XP_016882197.1
5
2994
GenBank mRNA
AB013897.1
-
5
2520
AK056243.1
-
6
2607
AK128116.1
-
9
3226
AK223616.1
-
5
2512
AK301601.1
-
5
1968
BC004513.2
-
1
1694
BC017256.2
-
6
2673
3356680
BC053845.1
-
4
2510
5493685
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
zinc finger transcription factor
C2H2 zinc finger transcription factor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-cq5mm:80/10.239.166.46:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0