Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF677 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317998.1 | NP_001304927.1 | 4 | 515 | ||
NM_182609.3 | NP_872415.1 | 4 | 333 | |||
XM_005258862.4 | XP_005258919.1 | 4 | 515 | |||
XM_011526912.2 | XP_011525214.1 | 4 | 461 | |||
GenBank mRNA | AK026366.1 | - | 4 | 496 | ||
AK308120.1 | - | 4 | 295 | |||
BC029855.1 | - | 4 | 322 | |||
BC050038.1 | - | 4 | 288 | 5272569 | ||
BX648243.1 | - | 4 | 304 | |||
CK125988.1 | - | 4 | 519 | |||
DA337379.1 | - | 4 | 515 | |||
DA592734.1 | - | 4 | 333 |