Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002455.4 | NP_002446.3 | 3 | 747 | ||
NM_198883.3 | NP_942584.2 | 3 | 747 | |||
GenBank mRNA | AK313863.1 | - | 4 | 682 | ||
AX775989.1 | - | 3 | 194 | |||
BC001906.1 | - | 3 | 649 | 3546178 | ||
BE394487.1 | - | 3 | 493 |