Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V1E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039366.1 | NP_001034455.1 | 8 | 769 | ||
NM_001039367.1 | NP_001034456.1 | 8 | 745 | |||
NM_001696.3 | NP_001687.1 | 9 | 835 | |||
GenBank mRNA | AK294623.1 | - | 8 | 696 | ||
AK312394.1 | - | 9 | 756 | |||
AK315941.1 | - | 8 | 671 | |||
AW406069.1 | - | 2 | 62 | |||
BC004443.1 | - | 9 | 737 | 2820930 | ||
BQ888820.1 | - | 8 | 715 | |||
BT007128.1 | - | 9 | 648 | |||
CR456385.1 | - | 9 | 701 | |||
X71491.1 | - | 9 | 704 | |||
X76228.1 | - | 9 | 723 |