Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002028.3 | NP_002019.1 | 1, 2 | 304 | ||
GenBank mRNA | AK225917.1 | - | 1, 2 | 171 | ||
AK315714.1 | - | 1, 2 | 371 | |||
AU122768.1 | - | 1, 2 | 304 | |||
BC020232.1 | - | 1, 2 | 154 | 4652202 | ||
BX248269.1 | - | 1, 2 | 150 | |||
L00635.1 | - | 1, 2 | 158 |