Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001118890.1 | NP_001112362.1 | 1 | 371 | ||
NM_001243658.1 | NP_001230587.1 | 1 | 371 | |||
NM_001243659.1 | NP_001230588.1 | 1 | 371 | |||
NM_002064.2 | NP_002055.1 | 1 | 371 | |||
GenBank mRNA | AA485178.1 | - | 1 | 164 | ||
AF069668.1 | - | 1 | 175 | |||
AF162769.1 | - | 1 | 163 | |||
AK311868.1 | - | 1 | 220 | |||
BC005304.1 | - | 1 | 224 | 3682161 | ||
BC010965.1 | - | 1 | 228 | 4333310 | ||
BC106075.1 | - | 1 | 227 | 4444172 | ||
BG547098.1 | - | 1 | 225 | |||
BP196332.1 | - | 1 | 371 | |||
BT006689.1 | - | 1 | 160 | |||
BU949348.1 | - | 1 | 147 | |||
BX106369.1 | - | 1 | 165 | |||
CR450312.1 | - | 1 | 160 | |||
CR542165.1 | - | 1 | 160 | |||
D21238.1 | - | 1 | 163 | |||
X76648.1 | - | 1 | 223 |