Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM7SF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277233.1 | NP_001264162.1 | 5 | 704 | ||
NM_003273.3 | NP_003264.2 | 5 | 704 | |||
NR_102367.1 | 4 | 649 | ||||
GenBank mRNA | AF023676.1 | - | 3 | 1198 | ||
AF096304.1 | - | 5 | 668 | |||
AK290935.1 | - | 5 | 682 | |||
AK296441.1 | - | 4 | 625 | |||
BC009052.1 | - | 5 | 670 | 3874367 | ||
BC038353.1 | - | 5 | 674 | 3861907 | ||
BG771219.1 | - | 5 | 690 |