Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSTA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317783.1 | NP_001304712.1 | 3 | 266 | ||
NM_003313.3 | NP_003304.1 | 3 | 303 | |||
XM_005251051.3 | XP_005251108.2 | 3 | 238 | |||
XM_011517269.1 | XP_011515571.1 | 3 | 330 | |||
GenBank mRNA | AK096752.1 | - | 2 | 995 | ||
AK303125.1 | - | 3 | 248 | |||
AK313560.1 | - | 3 | 245 | |||
BC001941.1 | - | 3 | 239 | 2819332 | ||
BC093061.1 | - | 3 | 280 | 30370158 | ||
BG709420.1 | - | 3 | 274 | |||
U58766.1 | - | 2 | 277 |