Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FKBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135211.2 | NP_001128683.1 | 7 | 960 | ||
NM_001281304.1 | NP_001268233.1 | 6 | 961 | |||
NM_003602.4 | NP_003593.3 | 7 | 1051 | |||
XM_006716153.2 | XP_006716216.1 | 6 | 835 | |||
XM_017012741.1 | XP_016868230.1 | 7 | 940 | |||
XM_017012742.1 | XP_016868231.1 | 7 | 975 | |||
GenBank mRNA | AF038847.1 | - | 7 | 915 | ||
AF447060.1 | - | 6 | 918 | |||
AK302121.1 | - | 7 | 948 | |||
BC036817.1 | - | 7 | 923 | 5744865 |