Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OASL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261825.1 | NP_001248754.1 | 4 | 943 | ||
NM_003733.3 | NP_003724.1 | 6 | 1333 | |||
NM_198213.2 | NP_937856.1 | 5 | 1091 | |||
GenBank mRNA | AJ225089.1 | - | 6 | 1068 | ||
AK023956.1 | - | 6 | 1069 | |||
AK314419.1 | - | 6 | 1068 | |||
BC117408.1 | - | 6 | 1086 | 40125959 | ||
BC117410.1 | - | 6 | 1086 | 40125961 | ||
BM909951.1 | - | 4 | 536 | |||
JQ792168.1 | - | 4 | 672 |