Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPNMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005340.1 | NP_001005340.1 | 2 | 372 | ||
NM_002510.2 | NP_002501.1 | 2 | 372 | |||
XM_005249578.2 | XP_005249635.1 | 2 | 863 | |||
XM_017011676.1 | XP_016867165.1 | 2 | 863 | |||
XM_017011677.1 | XP_016867166.1 | 2 | 863 | |||
XM_017011678.1 | XP_016867167.1 | 2 | 863 | |||
GenBank mRNA | AF322909.1 | - | 2 | 270 | ||
AJ505015.1 | - | 2 | 284 | |||
AK292078.1 | - | 2 | 294 | |||
AK296779.1 | - | 2 | 294 | |||
AK297031.1 | - | 2 | 294 | |||
AK297051.1 | - | 2 | 303 | |||
AK304671.1 | - | 3 | 353 | |||
AK312279.1 | - | 2 | 301 | |||
AY359124.1 | - | 2 | 292 | |||
BC011595.1 | - | 2 | 260 | |||
BC032783.1 | - | 2 | 288 | 5177095 | ||
BM750843.1 | - | 2 | 382 | |||
BT007074.1 | - | 2 | 211 | |||
CD105306.1 | - | 2 | 360 | |||
X76534.1 | - | 2 | 302 |