Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185039.1 | NP_001171968.1 | 12 | 2154 | ||
NM_004035.6 | NP_004026.2 | 12 | 1980 | |||
NM_007292.5 | NP_009223.2 | 12 | 1980 | |||
XM_011524868.2 | XP_011523170.1 | 13 | 2139 | |||
XM_011524869.2 | XP_011523171.1 | 11 | 1817 | |||
GenBank mRNA | AK225897.1 | - | 12 | 1680 | ||
AK291793.1 | - | 12 | 1674 | |||
AK292965.1 | - | 12 | 1679 | |||
AK296409.1 | - | 12 | 1794 | |||
AK300447.1 | - | 13 | 1835 | |||
AK308218.1 | - | 11 | 1419 | |||
BC008767.2 | - | 12 | 1909 | 3051501 | ||
BC010425.1 | - | 12 | 1698 | 3357409 | ||
BX537380.1 | - | 12 | 1639 | |||
S69189.1 | - | 12 | 1671 | |||
U07866.1 | - | 12 | 1619 | |||
X71440.1 | - | 12 | 1632 |