Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042422.2 | NP_001035887.1 | 5 | 1300 | ||
NM_001042423.2 | NP_001035888.1 | 5 | 1304 | |||
NM_001206950.1 | NP_001193879.1 | 5 | 1316 | |||
NM_001206951.1 | NP_001193880.1 | 5 | 1278 | |||
NM_001206952.1 | NP_001193881.1 | 5 | 1245 | |||
NM_004207.3 | NP_004198.1 | 5 | 1272 | |||
XM_011523607.1 | XP_011521909.1 | 5 | 3193 | |||
GenBank mRNA | AK127319.1 | - | 4 | 3460 | ||
AK223040.1 | - | 5 | 1230 | |||
BC112267.1 | - | 4 | 1191 | 8327735 | ||
BC112269.1 | - | 4 | 1191 | 8327737 | ||
U81800.1 | - | 5 | 1212 |