Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004569.3 | NP_004560.1 | 4 | 814 | ||
XM_011536838.2 | XP_011535140.1 | 5 | 934 | |||
XM_017021371.1 | XP_016876860.1 | 5 | 935 | |||
XM_017021372.1 | XP_016876861.1 | 4 | 804 | |||
GenBank mRNA | BC004100.2 | - | 4 | 765 | 3936489 | |
BC071849.1 | - | 4 | 762 | 5935599 | ||
L19783.1 | - | 4 | 777 |