Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC23A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005116.5 | NP_005107.4 | 17 | 6599 | ||
NM_203327.1 | NP_976072.1 | 17 | 6617 | |||
XM_011529414.2 | XP_011527716.1 | 17 | 6604 | |||
XM_011529415.2 | XP_011527717.1 | 16 | 6425 | |||
XM_011529416.2 | XP_011527718.1 | 16 | 6734 | |||
XM_011529417.2 | XP_011527719.1 | 16 | 6481 | |||
XM_017028171.1 | XP_016883660.1 | 17 | 6638 | |||
XM_017028172.1 | XP_016883661.1 | 17 | 6642 | |||
XM_017028173.1 | XP_016883662.1 | 17 | 6622 | |||
XM_017028174.1 | XP_016883663.1 | 17 | 6517 | |||
XM_017028175.1 | XP_016883664.1 | 13 | 5953 | |||
XM_017028176.1 | XP_016883665.1 | 14 | 6271 | |||
XM_017028177.1 | XP_016883666.1 | 14 | 6258 | |||
XM_017028178.1 | XP_016883667.1 | 16 | 6524 | |||
XM_017028179.1 | XP_016883668.1 | 16 | 6511 | |||
XM_017028180.1 | XP_016883669.1 | 12 | 5835 | |||
GenBank mRNA | AB075494.1 | - | 1 | 1470 | ||
AK025668.1 | - | 2 | 2919 | |||
D87075.2 | - | 17 | 6599 |