Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: HMGCS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166107.1 | NP_001159579.1 | 2 | 312 | ||
NM_005518.3 | NP_005509.1 | 2 | 312 | |||
XM_011541313.1 | XP_011539615.1 | 2 | 505 | |||
GenBank mRNA | AK301594.1 | - | 2 | 282 | ||
AK302276.1 | - | 2 | 285 | |||
AK303777.1 | - | 2 | 285 | |||
AK313590.1 | - | 2 | 287 | |||
BC044217.1 | - | 2 | 265 | 5744956 | ||
BF964973.1 | - | 2 | 286 | |||
BP420631.1 | - | 2 | 321 | |||
CR456850.1 | - | 2 | 224 | |||
U12788.1 | - | 1 | 120 | |||
X83618.1 | - | 2 | 275 |