Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ODF2 SIRNA Assays ›
Gene Symbol:
ODF2
Gene Name:
outer dense fiber of sperm tails 2
Gene Aliases:
CT134, ODF2/1, ODF2/2, ODF84
Chromosome Location:
Chr.9: 128455155 - 128501292 on Build GRCh38
UniGene ID:
Hs.129055
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
11735
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ODF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242352.1
NP_001229281.1
5, 6
800
NM_001242353.1
NP_001229282.1
5, 6
886
NM_002540.4
NP_002531.3
7, 8
1088
NM_153432.1
NP_702910.1
4, 5
646
NM_153433.1
NP_702911.1
5, 6
813
NM_153435.1
NP_702913.1
5, 6
706
NM_153436.1
NP_702914.1
4, 5
470
NM_153437.2
NP_702915.1
5, 6
736
NM_153439.1
NP_702917.1
4, 5
589
NM_153440.1
NP_702918.1
4, 5
516
XM_005252010.2
XP_005252067.1
4, 5
598
XM_005252011.2
XP_005252068.1
4, 5
538
XM_005252012.3
XP_005252069.1
5, 6
788
XM_005252013.3
XP_005252070.1
5, 6
735
XM_005252014.3
XP_005252071.1
5, 6
784
XM_005252017.4
XP_005252074.1
5, 6
689
XM_005252018.2
XP_005252075.1
6, 7
894
XM_005252019.3
XP_005252076.1
6, 7
913
XM_005252022.2
XP_005252079.1
5, 6
712
XM_005252023.2
XP_005252080.1
5, 6
683
XM_005252024.3
XP_005252081.1
5, 6
702
XM_005252026.4
XP_005252083.1
5, 6
631
XM_005252029.2
XP_005252086.1
5, 6
712
XM_006717126.3
XP_006717189.1
5, 6
788
XM_011518722.2
XP_011517024.1
5, 6
712
XM_011518723.2
XP_011517025.1
5, 6
655
XM_011518724.2
XP_011517026.1
6, 7
876
XM_011518725.2
XP_011517027.1
6, 7
880
XM_011518726.2
XP_011517028.1
6, 7
898
XM_011518728.2
XP_011517030.1
5, 6
649
XM_011518729.2
XP_011517031.1
5, 6
585
XM_011518730.2
XP_011517032.1
5, 6
712
XM_011518731.2
XP_011517033.1
4, 5
541
XM_011518732.2
XP_011517034.1
5, 6
498
XM_011518744.1
XP_011517046.1
4, 5
538
XM_017014764.1
XP_016870253.1
6, 7
869
XM_017014765.1
XP_016870254.1
6, 7
888
XM_017014766.1
XP_016870255.1
4, 5
598
XM_017014767.1
XP_016870256.1
6, 7
880
XM_017014768.1
XP_016870257.1
5, 6
672
XM_017014769.1
XP_016870258.1
5, 6
701
XM_017014770.1
XP_016870259.1
6, 7
856
XM_017014771.1
XP_016870260.1
6, 7
837
XM_017014772.1
XP_016870261.1
6, 7
913
XM_017014773.1
XP_016870262.1
6, 7
889
XM_017014774.1
XP_016870263.1
5, 6
599
XM_017014775.1
XP_016870264.1
6, 7
856
XM_017014776.1
XP_016870265.1
6, 7
837
XM_017014777.1
XP_016870266.1
5, 6
639
XM_017014778.1
XP_016870267.1
5, 6
742
XM_017014779.1
XP_016870268.1
4, 5
483
GenBank mRNA
AF012549.1
-
5, 6
676
AF053970.1
-
3, 4
375
AK093367.1
-
4, 5
596
AK299303.1
-
5, 6
648
AK301754.1
-
4, 5
513
AK301842.1
-
4, 5
438
AK301855.1
-
4, 5
492
AK302684.1
-
5, 6
740
AK308278.1
-
3, 4
352
AK309917.1
-
5, 6
591
AY319414.1
-
4, 5
622
AY366499.1
-
5, 6
583
BC010629.1
-
7, 8
886
3874501
BC091500.1
-
5, 6
700
30378678
BM563800.1
-
570
DC397869.1
-
4, 5
470
DQ444713.1
-
3, 4
413
DQ444714.1
-
3, 4
356
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
multicellular organism development
spermatid development
structural molecule activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0