Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098486.1 | NP_001091956.1 | 8 | 983 | ||
NM_006632.3 | NP_006623.2 | 7 | 749 | |||
GenBank mRNA | AK296070.1 | - | 5 | 845 | ||
AK296082.1 | - | 6 | 1077 | |||
AK298271.1 | - | 8 | 983 | |||
AK298297.1 | - | 5 | 791 | |||
AK298367.1 | - | 7 | 746 | |||
BC017952.1 | - | 7 | 718 | 4557795 | ||
CB163247.1 | - | 2 | 121 | |||
U90545.1 | - | 7 | 958 |