Gene Symbol: TXNL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303471.2 | NP_001290400.1 | 4 | 1409 | ||
NM_001305557.1 | NP_001292486.1 | 3 | 1236 | |||
NM_001305563.1 | NP_001292492.1 | 4 | 1049 | |||
NM_001305564.1 | NP_001292493.1 | 3 | 992 | |||
NM_006701.4 | NP_006692.1 | 3 | 1260 | |||
NR_131175.1 | 5 | 1479 | ||||
NR_131176.1 | 4 | 1330 | ||||
NR_131177.1 | 2 | 1156 | ||||
GenBank mRNA | AK308703.1 | - | 3 | 972 | ||
BC001046.2 | - | 3 | 1200 | 2823293 | ||
BC019272.1 | - | 3 | 1200 | 2823293 |