Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020474.3 | NP_065207.2 | 6 | 982 | ||
XM_005258239.3 | XP_005258296.1 | 7 | 1050 | |||
XM_017025692.1 | XP_016881181.1 | 7 | 1229 | |||
GenBank mRNA | AK293062.1 | - | 8 | 1124 | ||
BC038440.1 | - | 7 | 1047 | 4478643 | ||
U41514.1 | - | 6 | 919 | |||
X85018.1 | - | 6 | 888 |