Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134771.1 | NP_001128243.1 | 26 | 4147 | ||
NM_020708.4 | NP_065759.1 | 26 | 4165 | |||
XM_017027981.1 | XP_016883470.1 | 25 | 4132 | |||
GenBank mRNA | AF208159.1 | - | 26 | 4002 | ||
AK055066.1 | - | 1 | 132 | |||
BC036329.1 | - | 1 | 31 | |||
BC132668.1 | - | 26 | 4090 | 40146690 | ||
BC132670.1 | - | 26 | 4090 | 40146692 | ||
KC295697.1 | - | 3 | 391 |