Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GPAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244949.1 | NP_001231878.1 | 12 | 1372 | ||
NM_020918.5 | NP_065969.3 | 12 | 1368 | |||
XM_005269998.1 | XP_005270055.1 | 12 | 1513 | |||
GenBank mRNA | AB046780.1 | - | 6 | 660 | ||
AL832464.1 | - | 12 | 1324 | |||
AL833061.2 | - | 12 | 1354 | |||
AL833093.2 | - | 12 | 1357 | |||
BC030783.1 | - | 12 | 1336 | 4821342 | ||
BX647451.1 | - | 12 | 1336 |