Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037290.2 | NP_001032367.2 | 2 | 278 | ||
NM_001256397.1 | NP_001243326.1 | 2 | 278 | |||
NM_001256398.1 | NP_001243327.1 | 2 | 390 | |||
NM_031938.5 | NP_114144.4 | 2 | 390 | |||
GenBank mRNA | AB096252.1 | - | 2 | 244 | ||
AF276432.1 | - | 2 | 887 | |||
AJ290393.1 | - | 2 | 240 | |||
AK027801.1 | - | 2 | 273 | |||
AK315300.1 | - | 2 | 382 | |||
BC041656.1 | - | 2 | 353 | 5175925 | ||
BX503569.1 | - | 2 | 278 |