Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC5A11 SIRNA Assays ›
Gene Symbol:
SLC5A11
Gene Name:
solute carrier family 5 member 11
Gene Aliases:
KST1, RKST1, SGLT6, SMIT2
Chromosome Location:
Chr.16: 24832784 - 24911711 on Build GRCh38
UniGene ID:
Hs.164118
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
117709
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC5A11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001258411.2
NP_001245340.1
13, 14
1708
NM_001258412.2
NP_001245341.1
12, 13
1603
NM_001258413.2
NP_001245342.1
13, 14
1654
NM_001258414.1
NP_001245343.1
12, 13
1378
NM_052944.4
NP_443176.2
13, 14
1664
XM_005255080.2
XP_005255137.1
14, 15
1692
XM_005255082.4
XP_005255139.1
8, 9
1098
XM_006721015.2
XP_006721078.1
12, 13
1672
XM_011545725.2
XP_011544027.2
13, 14
1835
XM_011545728.1
XP_011544030.1
5, 6
631
XM_017022900.1
XP_016878389.1
14, 15
2008
XM_017022901.1
XP_016878390.1
14, 15
2718
XM_017022902.1
XP_016878391.1
14, 15
1847
XM_017022903.1
XP_016878392.1
15, 16
1857
XM_017022904.1
XP_016878393.1
15, 16
2027
XM_017022905.1
XP_016878394.1
13, 14
1903
XM_017022906.1
XP_016878395.1
14, 15
1868
XM_017022907.1
XP_016878396.1
13, 14
1416
XM_017022908.1
XP_016878397.1
13, 14
1475
XM_017022909.1
XP_016878398.1
12, 13
1311
GenBank mRNA
AF292385.1
-
12, 13
1432
AJ305237.1
-
12, 13
1450
AK125267.1
-
15, 16
1780
AK295427.1
-
12, 13
1472
AK298373.1
-
12, 13
1449
AK303691.1
-
13, 14
1576
AY044906.1
-
13, 14
1664
BC049385.1
-
12, 13
1248
5755223
BC057780.1
-
13, 14
1523
5287366
Back To Top
More Information
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Function(s)
Process(es)
sodium ion transport
apoptotic process
carbohydrate transport
polyol transport
transmembrane transport
protein binding
polyol transmembrane transporter activity
symporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-wh4bk:80/10.239.163.60:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0