Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123226.1 | NP_001116698.1 | 4 | 897 | ||
NM_012123.3 | NP_036255.2 | 4 | 897 | |||
NM_133645.2 | NP_598400.1 | 4 | 897 | |||
XM_006715444.2 | XP_006715507.1 | 4 | 794 | |||
XM_006715445.2 | XP_006715508.1 | 4 | 599 | |||
XM_006715446.3 | XP_006715509.1 | 4 | 897 | |||
XM_011535723.1 | XP_011534025.1 | 5 | 986 | |||
XM_011535724.1 | XP_011534026.1 | 5 | 854 | |||
XM_011535725.2 | XP_011534027.1 | 5 | 722 | |||
XM_017010697.1 | XP_016866186.1 | 4 | 794 | |||
XM_017010698.1 | XP_016866187.1 | 5 | 986 | |||
GenBank mRNA | AF132937.1 | - | 4 | 819 | ||
AF319422.2 | - | 4 | 863 | |||
AF469111.1 | - | 4 | 863 | |||
AK074625.1 | - | 4 | 876 | |||
AK225828.1 | - | 4 | 881 | |||
AL833823.1 | - | 4 | 741 | |||
AY078985.1 | - | 4 | 863 | |||
AY078986.1 | - | 4 | 863 | |||
BC005808.2 | - | 4 | 582 | |||
BC011051.1 | - | 4 | 838 | 4339769 |