Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138801.2 | NP_620156.1 | 2 | 464 | ||
XM_011532540.1 | XP_011530842.1 | 2 | 464 | |||
GenBank mRNA | AK291489.1 | - | 2 | 272 | ||
BC014916.1 | - | 2 | 246 | 3920640 | ||
BC019263.1 | - | 2 | 277 | 3502667 | ||
DA984992.1 | - | 2 | 464 | |||
EU794611.1 | - | 2 | 246 | |||
U11036.1 | - | 2 | 263 |