Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004204.3 | NP_004195.2 | 6, 7 | 1369 | ||
NM_148920.2 | NP_683721.1 | 6, 7 | 1369 | |||
GenBank mRNA | AB003723.1 | - | 6, 7 | 1333 | ||
AF030177.1 | - | 6, 7 | 1302 | |||
AK055163.1 | - | 20 | ||||
AK300297.1 | - | 6, 7 | 1336 | |||
AK314361.1 | - | 7, 8 | 1398 | |||
BC006377.2 | - | 6, 7 | 1319 | 4299132 |