Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318876.1 | NP_001305805.1 | 2, 3 | 223 | ||
NM_203290.3 | NP_976035.1 | 2, 3 | 223 | |||
XM_011515000.2 | XP_011513302.1 | 2, 3 | 164 | |||
GenBank mRNA | AF008442.1 | - | 2, 3 | 149 | ||
AF047441.1 | - | 2, 3 | 159 | |||
AK293561.1 | - | 2, 3 | 149 | |||
BC008118.1 | - | 2, 3 | 114 | |||
BC008863.2 | - | 2, 3 | 149 | 4128104 | ||
BM828768.1 | - | 2, 3 | 214 | |||
DW415914.1 | - | 2, 3 | 242 |