Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004878.4 | NP_004869.1 | 3 | 346 | ||
XM_011519210.2 | XP_011517512.1 | 4 | 707 | |||
GenBank mRNA | AF010316.1 | - | 3 | 305 | ||
AF027740.1 | - | 3 | 311 | |||
AF217965.1 | - | 4 | 418 | |||
AK127663.1 | - | 1 | 1488 | |||
AK311947.1 | - | 3 | 326 | |||
BC008280.1 | - | 3 | 328 | 3623516 | ||
BM550187.1 | - | 3 | 282 | |||
BQ927197.1 | - | 3 | 346 | |||
BX357872.2 | - | 3 | 330 | |||
BX396892.2 | - | 4 | 422 | |||
CB960064.1 | - | 3 | 384 |