Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PYGM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164716.1 | NP_001158188.1 | 12 | 2268 | ||
GenBank mRNA | AF066859.1 | - | 14 | 1715 | ||
AK056607.1 | - | 12 | 1527 | |||
AK299376.1 | - | 10 | 1055 | |||
AK299461.1 | - | 14 | 1734 | |||
AK314474.1 | - | 14 | 1719 | |||
BC126392.1 | - | 14 | 2114 | 8992108 | ||
BC130514.1 | - | 14 | 2114 | 40146545 | ||
M16013.1 | - | 4 | 353 |