Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130112.2 | NP_001123584.1 | 9, 10 | 1040 | ||
NM_007370.5 | NP_031396.1 | 8, 9 | 914 | |||
NM_181578.3 | NP_853556.2 | 9, 10 | 1278 | |||
XM_011538643.2 | XP_011536945.1 | 8, 9 | 912 | |||
XM_011538645.2 | XP_011536947.1 | 8, 9 | 912 | |||
XM_017019779.1 | XP_016875268.1 | 8, 9 | 912 | |||
GenBank mRNA | AK094575.1 | - | 9, 10 | 1257 | ||
AK291107.1 | - | 8, 9 | 898 | |||
BC001866.1 | - | 8, 9 | 888 | 3544137 | ||
BC013961.2 | - | 8, 9 | 872 | 2960023 | ||
BC020440.1 | - | 8, 9 | 845 | |||
CR407651.1 | - | 8, 9 | 787 | |||
L07540.1 | - | 8, 9 | 796 |