Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ECE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032331.3 | NP_115707.2 | 3 | 611 | ||
GenBank mRNA | BC005835.2 | - | 3 | 607 | ||
BC012449.1 | - | 3 | 584 | |||
BC069005.1 | - | 3 | 597 |