Gene Symbol: TXNL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303471.2 | NP_001290400.1 | 4 | 891 | ||
NM_001305557.1 | NP_001292486.1 | 3 | 718 | |||
NM_001305563.1 | NP_001292492.1 | 4 | 531 | |||
NM_001305564.1 | NP_001292493.1 | 3 | 474 | |||
NM_006701.4 | NP_006692.1 | 3 | 742 | |||
NR_131175.1 | 5 | 961 | ||||
NR_131176.1 | 4 | 812 | ||||
NR_131177.1 | 2 | 638 | ||||
GenBank mRNA | AF023611.1 | - | 3 | 668 | ||
AF023612.1 | - | 4 | 738 | |||
AK308703.1 | - | 3 | 454 | |||
AK309098.1 | - | 4 | 808 | |||
AV729855.2 | - | 3 | 474 | |||
BC001046.2 | - | 3 | 682 | 2823293 | ||
BC019272.1 | - | 3 | 682 | 2823293 | ||
BM922767.1 | - | 3 | 719 |