Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NRDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101662.1 | NP_001095132.1 | 12 | 1810 | ||
NM_001242361.1 | NP_001229290.1 | 14 | 1409 | |||
NM_002525.2 | NP_002516.2 | 14 | 2014 | |||
XM_005270903.1 | XP_005270960.1 | 12 | 1157 | |||
XM_011541522.1 | XP_011539824.1 | 15 | 2003 | |||
XM_011541523.1 | XP_011539825.1 | 12 | 1374 | |||
XM_011541525.2 | XP_011539827.1 | 14 | 1472 | |||
XM_017001375.1 | XP_016856864.1 | 12 | 1234 | |||
GenBank mRNA | AK055516.1 | - | 12 | 1154 | ||
AK299271.1 | - | 12 | 1257 | |||
AK302616.1 | - | 12 | 1234 | |||
AK311631.1 | - | 12 | 1204 | |||
AK312318.1 | - | 12 | 1622 | |||
AY049784.1 | - | 12 | 1525 | |||
AY360265.1 | - | 14 | 1385 | |||
BC008775.2 | - | 12 | 1584 | 3138712 | ||
U64898.1 | - | 12 | 1582 | |||
X93207.1 | - | 14 | 1827 | |||
X93209.1 | - | 12 | 1623 |