Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRKACB SIRNA Assays ›
Gene Symbol:
PRKACB
Gene Name:
protein kinase cAMP-activated catalytic subunit beta
Gene Aliases:
PKA C-beta, PKACB
Chromosome Location:
Chr.1: 84077975 - 84238498 on Build GRCh38
UniGene ID:
Hs.487325
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
118253
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRKACB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242857.2
NP_001229786.1
5
325
NM_001242858.2
NP_001229787.1
2
268
NM_001242859.2
NP_001229788.1
4
316
NM_001242860.2
NP_001229789.1
5
460
NM_001242861.2
NP_001229790.1
4
451
NM_001300915.1
NP_001287844.1
5
460
NM_001300916.1
NP_001287845.1
2
330
NM_001300917.1
NP_001287846.1
2
268
NM_002731.3
NP_002722.1
2
426
NM_182948.3
NP_891993.1
2
330
NM_207578.2
NP_997461.1
2
426
XM_005271015.3
XP_005271072.1
4
277
XM_005271016.1
XP_005271073.1
4
402
XM_005271017.3
XP_005271074.1
3
267
XM_005271018.4
XP_005271075.1
3
587
XM_005271019.1
XP_005271076.1
2
343
XM_005271020.1
XP_005271077.1
2
310
XM_006710758.1
XP_006710821.1
4
416
XM_017001706.1
XP_016857195.1
5
320
XM_017001707.1
XP_016857196.1
5
320
XM_017001708.1
XP_016857197.1
5
421
XM_017001709.1
XP_016857198.1
4
118
XM_017001710.1
XP_016857199.1
4
416
XM_017001711.1
XP_016857200.1
4
118
XM_017001712.1
XP_016857201.1
4
416
XM_017001713.1
XP_016857202.1
2
368
XM_017001714.1
XP_016857203.1
4
402
XM_017001715.1
XP_016857204.1
4
402
XM_017001716.1
XP_016857205.1
3
270
XM_017001717.1
XP_016857206.1
3
591
XM_017001718.1
XP_016857207.1
2
245
XM_017001719.1
XP_016857208.1
2
343
XM_017001720.1
XP_016857209.1
2
343
GenBank mRNA
AB209189.1
-
5
396
AB451239.1
-
2
75
AB451364.1
-
2
75
AK091420.1
-
4
125
AK296482.1
-
5
127
AK304322.1
-
2
255
AK304375.1
-
2
70
AK314550.1
-
2
322
AY927364.1
-
2
266
AY927365.1
-
2
120
AY927366.1
-
2
150
AY927367.1
-
4
194
AY927368.1
-
5
203
BC016285.1
-
2
257
3916100
BC035058.1
-
2
344
5262372
BI598037.1
-
2
140
BM905428.1
-
2
212
BX485407.1
-
2
311
BX537705.1
-
2
307
BX641026.1
-
2
306
BX647515.1
-
5
405
CR936631.1
-
4
119
DA188681.1
-
5
460
DC322547.1
-
5
326
HY160843.1
-
2
426
M34181.1
-
2
122
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
stimulatory C-type lectin receptor signaling pathway
renal water homeostasis
protein phosphorylation
signal transduction
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
blood coagulation
activation of protein kinase A activity
lipoprotein metabolic process
negative regulation of meiotic cell cycle
regulation of protein processing
cellular response to glucagon stimulus
response to clozapine
negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
magnesium ion binding
protein serine/threonine kinase activity
cAMP-dependent protein kinase activity
protein binding
ATP binding
kinase activity
ubiquitin protein ligase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0