Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAMK2D SIRNA Assays ›
Gene Symbol:
CAMK2D
Gene Name:
calcium/calmodulin dependent protein kinase II delta
Gene Aliases:
CAMKD
Chromosome Location:
Chr.4: 113451032 - 113761927 on Build GRCh38
UniGene ID:
Hs.144114
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
118257
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAMK2D
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001221.3
NP_001212.2
17
2092
NM_001321566.1
NP_001308495.1
18
2134
NM_001321567.1
NP_001308496.1
19
2194
NM_001321568.1
NP_001308497.1
19
2167
NM_001321569.1
NP_001308498.1
19
2194
NM_001321570.1
NP_001308499.1
19
2194
NM_001321571.1
NP_001308500.1
19
2194
NM_001321572.1
NP_001308501.1
19
2194
NM_001321573.1
NP_001308502.1
19
2167
NM_001321574.1
NP_001308503.1
17
2089
NM_001321575.1
NP_001308504.1
18
2134
NM_001321576.1
NP_001308505.1
17
2089
NM_001321577.1
NP_001308506.1
18
2131
NM_001321578.1
NP_001308507.1
17
2061
NM_001321579.1
NP_001308508.1
19
2194
NM_001321580.1
NP_001308509.1
18
2152
NM_001321581.1
NP_001308510.1
19
2235
NM_001321582.1
NP_001308511.1
18
2125
NM_001321583.1
NP_001308512.1
18
2134
NM_001321584.1
NP_001308513.1
18
2131
NM_001321585.1
NP_001308514.1
19
2228
NM_001321586.1
NP_001308515.1
19
2191
NM_001321587.1
NP_001308516.1
19
2167
NM_001321588.1
NP_001308517.1
18
2131
NM_001321589.1
NP_001308518.1
18
2134
NM_001321590.1
NP_001308519.1
18
2134
NM_001321591.1
NP_001308520.1
19
2167
NM_172114.1
NP_742112.1
18
2134
NM_172115.2
NP_742113.1
17
2092
NM_172127.2
NP_742125.1
17
2092
NM_172128.2
NP_742126.1
17
2092
NM_172129.1
NP_742127.1
18
2125
XM_005263253.3
XP_005263310.1
19
2009
XM_005263254.3
XP_005263311.1
19
2030
XM_006714330.2
XP_006714393.1
17
1931
XM_011532289.1
XP_011530591.1
18
1943
XM_011532290.1
XP_011530592.1
18
1940
XM_011532291.1
XP_011530593.1
18
1943
XM_011532292.2
XP_011530594.1
15
1044
XM_011532295.1
XP_011530597.1
18
1964
XM_017008672.1
XP_016864161.1
18
1973
XM_017008673.1
XP_016864162.1
18
1970
XM_017008674.1
XP_016864163.1
19
2030
XM_017008675.1
XP_016864164.1
20
2134
XM_017008676.1
XP_016864165.1
19
1370
XM_017008677.1
XP_016864166.1
18
1970
XM_017008678.1
XP_016864167.1
17
1907
GenBank mRNA
AB209288.1
-
18
1770
AF071569.1
-
17
1233
AK025368.1
-
6
343
AK055642.1
-
17
1610
AK294517.1
-
19
1485
AL120285.1
-
2
45
AY987011.1
-
18
1266
BC032784.1
-
17
1295
5178265
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
G1/S transition of mitotic cell cycle
MAPK cascade
regulation of cell growth
response to hypoxia
regulation of the force of heart contraction
regulation of membrane depolarization
regulation of transcription from RNA polymerase II promoter
protein phosphorylation
calcium ion transport
regulation of heart contraction
regulation of G2/M transition of mitotic cell cycle
positive regulation of cardiac muscle hypertrophy
regulation of cell communication by electrical coupling
positive regulation of cardiac muscle cell apoptotic process
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
positive regulation of smooth muscle cell migration
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
cellular potassium ion homeostasis
endoplasmic reticulum calcium ion homeostasis
positive regulation of Rac protein signal transduction
positive regulation of GTPase activity
protein autophosphorylation
positive regulation of smooth muscle cell proliferation
protein oligomerization
relaxation of cardiac muscle
regulation of ryanodine-sensitive calcium-release channel activity
interferon-gamma-mediated signaling pathway
regulation of cellular localization
cell growth involved in cardiac muscle cell development
positive regulation of ERK1 and ERK2 cascade
cellular response to calcium ion
cardiac muscle cell contraction
regulation of heart rate by cardiac conduction
regulation of cardiac muscle cell action potential
regulation of cardiac muscle cell action potential involved in regulation of contraction
regulation of cellular response to heat
regulation of histone deacetylase activity
regulation of cell communication by electrical coupling involved in cardiac conduction
regulation of relaxation of cardiac muscle
negative regulation of sodium ion transmembrane transport
regulation of calcium ion transmembrane transport via high voltage-gated calcium channel
negative regulation of sodium ion transmembrane transporter activity
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
Ras guanyl-nucleotide exchange factor activity
protein binding
calmodulin binding
ATP binding
sodium channel inhibitor activity
titin binding
protein homodimerization activity
ion channel binding
nitric-oxide synthase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0