Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113182.2 | NP_001106653.1 | 3 | 1835 | ||
NM_001199455.1 | NP_001186384.1 | 2 | 1525 | |||
NM_001199456.1 | NP_001186385.1 | 2 | 238 | |||
NM_001291986.1 | NP_001278915.1 | 3 | 1835 | |||
NM_005104.3 | NP_005095.1 | 3 | 1922 | |||
GenBank mRNA | AK056504.1 | - | 3 | 1793 | ||
AL832722.1 | - | 3 | 1770 | |||
BC000477.2 | - | 2 | 894 | |||
BC063840.1 | - | 2 | 390 | 6181728 | ||
BX647233.1 | - | 2 | 238 | |||
BX648109.1 | - | 3 | 1836 | |||
D42040.1 | - | 3 | 1922 | |||
M80613.1 | - | 2 | 1257 | |||
S78771.1 | - | 1 | 209 | |||
X62083.1 | - | 2 | 1257 |