Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAOK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252043.1 | NP_001238972.1 | 14 | 2577 | ||
NM_004783.3 | NP_004774.1 | 14 | 2577 | |||
NM_016151.3 | NP_057235.2 | 14 | 2577 | |||
XM_011545982.1 | XP_011544284.1 | 14 | 2824 | |||
XM_011545983.2 | XP_011544285.1 | 14 | 2821 | |||
XM_011545984.1 | XP_011544286.1 | 14 | 2824 | |||
XM_011545985.1 | XP_011544287.1 | 14 | 2821 | |||
XM_011545986.2 | XP_011544288.1 | 14 | 2824 | |||
GenBank mRNA | AB020688.1 | - | 14 | 2163 | ||
AF061943.1 | - | 14 | 2518 | |||
AF263313.1 | - | 13 | 1760 | |||
AK291473.1 | - | 14 | 2094 | |||
AK304446.1 | - | 6 | 1083 | |||
AL137701.1 | - | 6 | 2414 | |||
BC015204.1 | - | 14 | 2543 | |||
BC031825.2 | - | 13 | 1328 | |||
BC051798.1 | - | 14 | 2334 | |||
BC054495.1 | - | 14 | 2530 | |||
BC136653.1 | - | 14 | 2262 | 9020643 | ||
BC136655.1 | - | 14 | 2262 | 9020645 | ||
BC142663.1 | - | 14 | 1917 | 40148112 | ||
BC144344.1 | - | 14 | 2262 | 9052864 | ||
BC151221.1 | - | 14 | 1917 | 40148752 | ||
BC152413.1 | - | 14 | 2163 | 8862518 |