Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNC1LI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016141.3 | NP_057225.2 | 13 | 1851 | ||
GenBank mRNA | AF078849.1 | - | 13 | 1828 | ||
AK074348.1 | - | 13 | 1835 | |||
AK293253.1 | - | 11 | 1489 | |||
BC131620.1 | - | 13 | 1797 | 40117391 | ||
BX649189.1 | - | 11 | 4264 |