Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYBPC1 SIRNA Assays ›
Gene Symbol:
MYBPC1
Gene Name:
myosin binding protein C, slow type
Gene Aliases:
LCCS4, MYBPCC, MYBPCS
Chromosome Location:
Chr.12: 101594849 - 101695841 on Build GRCh38
UniGene ID:
Hs.654589
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
118355
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MYBPC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001254718.1
NP_001241647.1
5
394
NM_001254719.1
NP_001241648.1
5
394
NM_001254720.1
NP_001241649.1
4
358
NM_001254721.1
NP_001241650.1
5
394
NM_001254722.1
NP_001241651.1
3
316
NM_001254723.1
NP_001241652.1
4
355
NM_002465.3
NP_002456.2
7
469
NM_206819.2
NP_996555.1
7
469
NM_206820.2
NP_996556.1
5
394
NM_206821.2
NP_996557.1
5
394
XM_005268876.4
XP_005268933.1
6
403
XM_006719405.3
XP_006719468.1
7
330
XM_006719406.3
XP_006719469.1
7
330
XM_006719407.3
XP_006719470.1
6
403
XM_006719408.3
XP_006719471.1
7
330
XM_006719409.3
XP_006719472.1
5
356
XM_006719410.3
XP_006719473.1
7
330
XM_006719411.3
XP_006719474.1
5
356
XM_017019315.1
XP_016874804.1
7
330
XM_017019316.1
XP_016874805.1
7
330
XM_017019317.1
XP_016874806.1
5
356
XM_017019318.1
XP_016874807.1
7
330
XM_017019319.1
XP_016874808.1
5
356
XM_017019320.1
XP_016874809.1
3
398
XM_017019321.1
XP_016874810.1
5
356
XM_017019322.1
XP_016874811.1
3
398
GenBank mRNA
AK056587.1
-
6
393
AK292976.1
-
7
429
AK295727.1
-
3
276
AK296681.1
-
5
358
AK303401.1
-
4
315
AL831993.2
-
7
404
AL832000.2
-
7
404
AL832332.2
-
7
404
AL832350.2
-
7
404
AL832354.2
-
7
406
AL832456.2
-
7
404
AL834249.2
-
7
404
BC015187.1
-
7
429
BC065720.1
-
3
279
BC092418.1
-
5
341
30529254
BC117217.1
-
3
251
40125768
BC143495.1
-
5
329
9052003
BC143502.1
-
3
251
9052010
BC143503.1
-
5
329
9052011
BC143504.1
-
4
293
9052012
BP232990.1
-
5
354
BX499532.1
-
7
432
BX649073.2
-
8
500
DA433791.1
-
5
358
X73114.1
-
5
335
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
muscle filament sliding
positive regulation of protein targeting to mitochondrion
actin binding
protein binding
structural constituent of muscle
titin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-9ljwx:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0