Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PALM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040134.1 | NP_001035224.1 | 5 | 601 | ||
NM_002579.2 | NP_002570.2 | 5 | 601 | |||
XM_005259565.4 | XP_005259622.1 | 5 | 617 | |||
XM_005259566.4 | XP_005259623.1 | 5 | 960 | |||
XM_017026850.1 | XP_016882339.1 | 5 | 640 | |||
GenBank mRNA | AK022919.1 | - | 2 | 277 | ||
AK299809.1 | - | 5 | 457 | |||
AK301181.1 | - | 2 | 217 | |||
AK315168.1 | - | 5 | 453 | |||
BC019596.1 | - | 4 | 432 | |||
BC032449.1 | - | 5 | 449 | 5179938 | ||
BU508077.1 | - | 600 | ||||
D87460.1 | - | 2 | 281 | |||
DA521203.1 | - | 5 | 427 | |||
Y14770.1 | - | 5 | 434 | |||
Y16278.1 | - | 4 | 407 |